Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Trifluoromethyl)cyclopentane-1-carboxylic acid, Thermo Scientific™

Catalog No. MAY00118DE
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAY00118DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00118DE Supplier Thermo Scientific Chemicals Supplier No. MAY00118DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 277756-44-2
Molecular Formula C7H9F3O2
Molecular Weight (g/mol) 182.142
InChI Key DGQRYPPBAJNZFZ-UHFFFAOYSA-N
Synonym 1-trifluoromethyl cyclopentane-1-carboxylic acid, 1-trifluoromethyl cyclopentanecarboxylic acid, acmc-1cp3t, 1-trifluoromethyl-cyclopentanecarboxylic acid, 1-trifluoromethylcyclopentane carboxylic acid, cyclopentanecarboxylicacid, 1-trifluoromethyl
PubChem CID 15507525
IUPAC Name 1-(trifluoromethyl)cyclopentane-1-carboxylic acid
SMILES C1CCC(C1)(C(=O)O)C(F)(F)F

Specifications

Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 182.14
Percent Purity 97%
Chemical Name or Material 1-(Trifluoromethyl)cyclopentane-1-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.