accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 2778-96-3

IUPAC Name: octadecyl octadecanoate
Synonyms: stearyl stearate
Molecular Weight (g/mol): 536.97
Molecular Formula: C36H72O2
InChi Key: NKBWPOSQERPBFI-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCCCC
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results