Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearyl stearate, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013803 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3803 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3803 Supplier MilliporeSigma Supelco Supplier No. 46408100MG
Add to Cart
Add to Cart

Stearyl stearate is the ester of stearic acid and 1-octadecanol.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2778-96-3
Physical Form Neat
Linear Formula CH3(CH2)16 COO(CH2)17 CH3
MDL Number MFCD00056225
Synonym Stearic acid stearyl ester
Type Analytical Standard
Quantity 100 mg
Formula Weight 536.96
Percent Purity ≥98% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.