accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27825-21-4

IUPAC Name: methyl 3-chloropyrazine-2-carboxylate
Synonyms: methyl 3-chloropyrazine-2-carboxylate
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: SSXOFWYPCZOGBM-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC=CN=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results