Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Chloropyrazine-2-carboxylate 98.0+%, TCI America™
SDP

Catalog No. M26481G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M26481G Supplier TCI America Supplier No. M26481G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27825-21-4
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD03093326
InChI Key SSXOFWYPCZOGBM-UHFFFAOYSA-N
Synonym methyl 3-chloro-2-pyrazinecarboxylate, 3-chloro-pyrazine-2-carboxylic acid methyl ester, methyl3-chloropyrazine-2-carboxylate, 3-chloropyrazine-2-carboxylic acid methyl ester, pyrazinecarboxylic acid, 3-chloro-, methyl ester, 3-chloro-2-pyrazinecarboxylic acid methyl ester, ksc201c2b, methyl3-chloro-2-pyrazinecarboxylate, methyl 2-chloropyrazine-3-carboxylate, methyl 2-chloro-pyridizanine-3-carboxylate
PubChem CID 1519497
IUPAC Name methyl 3-chloropyrazine-2-carboxylate
SMILES COC(=O)C1=NC=CN=C1Cl

Specifications

Melting Point 33°C
Color White-Yellow
Boiling Point 89°C
Quantity 1 g
Formula Weight 172.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 3-Chloropyrazine-2-carboxylate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.