CAS RN 27846-49-7
IUPAC Name:
ethyl 3-(acetyloxy)butanoate
Synonyms:
ethyl 3-(acetyloxy)butanoate
Molecular Weight (g/mol):
174.2
Molecular Formula:
C8H14O4
InChi Key:
JFWYJXYRFBRWSH-UHFFFAOYNA-N
SMILES:
CCOC(=O)CC(C)OC(C)=O
1
–
1
of
1
results
1
Ethyl DL-3-Acetoxybutyrate 98.0+%, TCI America™
CAS: 27846-49-7 Molecular Formula: C8H14O4 Molecular Weight (g/mol): 174.20 MDL Number: MFCD00059378 InChI Key: JFWYJXYRFBRWSH-UHFFFAOYNA-N Synonym: DL-3-Acetoxybutyric Acid Ethyl Ester PubChem CID: 537574 IUPAC Name: ethyl 3-(acetyloxy)butanoate SMILES: CCOC(=O)CC(C)OC(C)=O