Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl DL-3-Acetoxybutyrate 98.0+%, TCI America™
SDP

Catalog No. A08315ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08315ML 5 mL
A083125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08315ML Supplier TCI America Supplier No. A08315ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27846-49-7
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.20
MDL Number MFCD00059378
InChI Key JFWYJXYRFBRWSH-UHFFFAOYNA-N
Synonym DL-3-Acetoxybutyric Acid Ethyl Ester
PubChem CID 537574
IUPAC Name ethyl 3-(acetyloxy)butanoate
SMILES CCOC(=O)CC(C)OC(C)=O

Specifications

Color Yellow
Boiling Point 94°C
Quantity 5 mL
UN Number 3272
Formula Weight 174.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl DL-3-Acetoxybutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.