accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OOHOHO

CAS RN 27899-21-4

IUPAC Name: 2,4-diethylpentanedioic acid
Synonyms: 2,4-diethylpentanedioic acid
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: CEAKZVDHZILFGK-UHFFFAOYNA-N
SMILES: CCC(CC(CC)C(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results