Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diethylglutaric Acid (DL- and meso- mixture) 98.0+%, TCI America™
SDP

Catalog No. D32345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32345G 5 g
D323425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32345G Supplier TCI America Supplier No. D32345G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27899-21-4
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD04117827
InChI Key CEAKZVDHZILFGK-UHFFFAOYSA-N
Synonym 2,4-Diethylpentanedioic Acid
PubChem CID 11586419
IUPAC Name 2,4-diethylpentanedioic acid
SMILES CCC(CC(CC)C(=O)O)C(=O)O

Specifications

Melting Point 78°C
Color White-Yellow
Quantity 5 g
Formula Weight 188.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diethylglutaric Acid (DL- and meso- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.