accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 279-49-2

IUPAC Name: 7-oxabicyclo[2.2.1]heptane
Synonyms: 7-oxabicyclo[2.2.1]heptane
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: YPWFNLSXQIGJCK-UHFFFAOYSA-N
SMILES: C1CC2CCC1O2
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)