accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 279261-89-1

IUPAC Name: (3,4-dihydro-2H-1,5-benzodioxepin-7-yl)boronic acid
Synonyms: 3,4-dihydro-2H-1,5-benzodioxepin-7-ylboronic acid
Molecular Weight (g/mol): 193.99
Molecular Formula: C9H11BO4
InChi Key: CSNCBPVLUIFJOS-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C2OCCCOC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results