accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3ClCl

CAS RN 27986-38-5

IUPAC Name: methyl 2,2-dichlorooctadecanoate
Synonyms: methyl 2,2-dichlorooctadecanoate
Molecular Weight (g/mol): 367.4
Molecular Formula: C19H36Cl2O2
InChi Key: HDINWBRYKNUCEC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCC(Cl)(Cl)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results