Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Dichlorostearate (mixture of isomers), TCI America™
SDP

Catalog No. D099925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D099925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D099925G Supplier TCI America Supplier No. D099925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27986-38-5
Molecular Formula C19H36Cl2O2
Molecular Weight (g/mol) 367.395
MDL Number MFCD00060077
InChI Key HDINWBRYKNUCEC-UHFFFAOYSA-N
Synonym Dichlorooctadecanoic Acid Methyl Ester, Dichlorostearic Acid Methyl Ester, Methyl Dichlorooctadecanoate
PubChem CID 53871329
IUPAC Name methyl 2,2-dichlorooctadecanoate
SMILES CCCCCCCCCCCCCCCCC(C(=O)OC)(Cl)Cl

Specifications

Color Yellow
Quantity 25 g
Formula Weight 367.40
Physical Form Liquid
Chemical Name or Material Methyl Dichlorostearate (mixture of isomers)
EINECSNumber (2)-1175&(2)-1181
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.