accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2799-21-5

IUPAC Name: (3R)-3-hydroxypyrrolidin-1-ium
Synonyms: (3R)-3-hydroxypyrrolidin-1-ium
Molecular Weight (g/mol): 88.13
Molecular Formula: C4H10NO
InChi Key: JHHZLHWJQPUNKB-SCSAIBSYSA-O
SMILES: O[C@@H]1CC[NH2+]C1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)