CAS RN 280-57-9
1,4-Diazabicyclo[2.2.2]octane, 98%, Thermo Scientific Chemicals
CAS: 280-57-9 Molecular Formula: C6H12N2 Molecular Weight (g/mol): 112.176 MDL Number: MFCD00006689 InChI Key: IMNIMPAHZVJRPE-UHFFFAOYSA-N Synonym: 1,4-diazabicyclo 2.2.2 octane,triethylenediamine,dabco,dabco 33lv,1,4-ethylenepiperazine,dabco crystal,teda,texacat td 100,dabco s-25 PubChem CID: 9237 IUPAC Name: 1,4-diazabicyclo[2.2.2]octane SMILES: C1CN2CCN1CC2
1,4-Diazabicyclo[2.2.2]octane 98.0+%, TCI America™
CAS: 280-57-9 Molecular Formula: C6H12N2 Molecular Weight (g/mol): 112.176 MDL Number: MFCD00006689 InChI Key: IMNIMPAHZVJRPE-UHFFFAOYSA-N Synonym: 1,4-diazabicyclo 2.2.2 octane,triethylenediamine,dabco,dabco 33lv,1,4-ethylenepiperazine,dabco crystal,teda,texacat td 100,dabco s-25 PubChem CID: 9237 IUPAC Name: 1,4-diazabicyclo[2.2.2]octane SMILES: C1CN2CCN1CC2
1,4-Diazabicyclo[2.2.2]octane, 97%
CAS: 280-57-9 Molecular Formula: C6H12N2 Molecular Weight (g/mol): 112.17 MDL Number: MFCD00006689 InChI Key: IMNIMPAHZVJRPE-UHFFFAOYSA-N Synonym: 1,4-diazabicyclo 2.2.2 octane,triethylenediamine,dabco,dabco 33lv,1,4-ethylenepiperazine,dabco crystal,teda,texacat td 100,dabco s-25 PubChem CID: 9237 IUPAC Name: 1,4-diazabicyclo[2.2.2]octane SMILES: C1CN2CCN1CC2
Sigma Aldrich 1,4-Diazabicyclo[2.2.2]Octane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Dabco™ 33-LV
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 2,2'-Bipyridyl
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc 1,4-Diazabicyclo-[2.2.2]-Octane, MP Biomedicals
CAS: 280-57-9 Molecular Formula: C6H12N2 Molecular Weight (g/mol): 112.176 MDL Number: MFCD00006689 InChI Key: IMNIMPAHZVJRPE-UHFFFAOYSA-N Synonym: 1,4-diazabicyclo 2.2.2 octane,triethylenediamine,dabco,dabco 33lv,1,4-ethylenepiperazine,dabco crystal,teda,texacat td 100,dabco s-25 PubChem CID: 9237 IUPAC Name: 1,4-diazabicyclo[2.2.2]octane SMILES: C1CN2CCN1CC2