Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diazabicyclo[2.2.2]octane 98.0+%, TCI America™
SDP

Catalog No. D0134100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0134100G 100 g
D013425G 25 g
D0134500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0134100G Supplier TCI America Supplier No. D0134100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 280-57-9
Molecular Formula C6H12N2
Molecular Weight (g/mol) 112.176
MDL Number MFCD00006689
InChI Key IMNIMPAHZVJRPE-UHFFFAOYSA-N
Synonym 1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25
PubChem CID 9237
IUPAC Name 1,4-diazabicyclo[2.2.2]octane
SMILES C1CN2CCN1CC2

Specifications

Melting Point 155°C
Color White-Yellow
Boiling Point 174°C
Quantity 100 g
UN Number 3263
Formula Weight 112.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diazabicyclo[2.2.2]octane
EINECSNumber (5)-1141
RTECSNumber HM0354200
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.