accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2805-56-3

IUPAC Name: (2E)-1,3-bis(4-fluorophenyl)prop-2-en-1-one
Synonyms: (2E)-1,3-bis(4-fluorophenyl)prop-2-en-1-one
Molecular Weight (g/mol): 244.24
Molecular Formula: C15H10F2O
InChi Key: AZBVDMNDUOURGI-XCVCLJGOSA-N
SMILES: FC1=CC=C(\C=C\C(=O)C2=CC=C(F)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results