Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Difluorochalcone 97.0+%, TCI America™
SDP

Catalog No. D28845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28845G 5 g
D288425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28845G Supplier TCI America Supplier No. D28845G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2805-56-3
Molecular Formula C15H10F2O
Molecular Weight (g/mol) 244.24
MDL Number MFCD00219906
InChI Key AZBVDMNDUOURGI-XCVCLJGOSA-N
Synonym 4,4'-difluorochalcone, e-1,3-bis 4-fluorophenyl prop-2-en-1-one, 2e-1,3-bis 4-fluorophenyl prop-2-en-1-one, 1,3-bis 4-fluorophenyl prop-2-en-1-one, trans-4,4'-difluorochalcone, 2-propen-1-one,1,3-bis 4-fluorophenyl, 4,4'-difluoro chalcone, 4,4/'-difluorochalcone, e-4,4'-difluorochalcone, azbvdmnduourgi-xcvcljgosa
PubChem CID 5286094
IUPAC Name (2E)-1,3-bis(4-fluorophenyl)prop-2-en-1-one
SMILES FC1=CC=C(\C=C\C(=O)C2=CC=C(F)C=C2)C=C1

Specifications

Melting Point 119°C
Color Yellow
Quantity 5 g
Formula Weight 244.24
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Difluorochalcone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.