accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28056-54-4

IUPAC Name: (4S)-2,2,4-trimethylcyclopentan-1-one
Synonyms: (4S)-2,2,4-trimethylcyclopentan-1-one
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: PJXIBUIXCXSNCM-ZCFIWIBFSA-N
SMILES: C[C@@H]1CC(=O)C(C)(C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results