Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4-Trimethylcyclopentanone 95.0+%, TCI America™
SDP

Catalog No. T17195ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17195ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17195ML Supplier TCI America Supplier No. T17195ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28056-54-4
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00019312
InChI Key PJXIBUIXCXSNCM-UHFFFAOYSA-N
PubChem CID 119632
IUPAC Name 2,2,4-trimethylcyclopentan-1-one
SMILES CC1CC(=O)C(C1)(C)C

Specifications

Color Yellow
Boiling Point 66°C
Quantity 5 mL
UN Number 1224
Formula Weight 126.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,4-Trimethylcyclopentanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.