accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2806-97-5

IUPAC Name: 1,1-diethoxybut-2-yne
Synonyms: 1,1-diethoxybut-2-yne
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: FKKVKKSEVMQYER-UHFFFAOYSA-N
SMILES: CCOC(OCC)C#CC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)