accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOHOH(Z)

CAS RN 28063-42-5

IUPAC Name: 1,3-dihydroxypropan-2-yl (13Z)-docos-13-enoate
Synonyms: glyceryl erucate
Molecular Weight (g/mol): 412.66
Molecular Formula: C25H48O4
InChi Key: TZDOBCZHGBGJLS-KTKRTIGZSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCCCCCC(=O)OC(CO)CO
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results