Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monoerucin, TCI America™
SDP

Catalog No. M10761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10761G Supplier TCI America Supplier No. M10761G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28063-42-5
Molecular Formula C25H48O4
Molecular Weight (g/mol) 412.655
MDL Number MFCD00063456
InChI Key ZXNAIPHYBVMMPY-MDZDMXLPSA-N
Synonym Glycerol alpha-Monoerucate, 1-Mono-cis-13-docosenoyl Glycerol
PubChem CID 6537151
IUPAC Name 2,3-dihydroxypropyl (E)-docos-13-enoate
SMILES CCCCCCCCC=CCCCCCCCCCCCC(=O)OCC(CO)O

Specifications

Melting Point 52°C
Color White
Quantity 1 g
Formula Weight 412.66
Physical Form Crystalline Powder
Chemical Name or Material Monoerucin
EINECSNumber (2)-0669
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.