accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3OCH3(E)

CAS RN 28069-74-1

IUPAC Name: 1-ethoxy-1-[(3E)-hex-3-en-1-yloxy]ethane
Synonyms: acetaldehyde ethyl hexenyl acetal
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: PAEBAEDUARAOSG-VOTSOKGWNA-N
SMILES: CCOC(C)OCC\C=C\CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results