accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28086-43-3

OHOHOOPHOHOONOHNO

CAS RN 28086-43-3

IUPAC Name: {[5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
Synonyms: uridine monophosphate
Molecular Weight (g/mol): 324.18
Molecular Formula: C9H13N2O9P
InChi Key: DJJCXFVJDGTHFX-UHFFFAOYNA-N
SMILES: OC1C(O)C(OC1COP(O)(O)=O)N1C=CC(=O)NC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results