accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2809-69-0

IUPAC Name: hexa-2,4-diyne
Synonyms: 2,4-hexadiyne
Molecular Weight (g/mol): 78.11
Molecular Formula: C6H6
InChi Key: PCTCNWZFDASPLA-UHFFFAOYSA-N
SMILES: CC#CC#CC
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)