Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Hexadiyne, 98%, Thermo Scientific™

Catalog No. AAL0637709
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0637709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0637709 Supplier Thermo Scientific Chemicals Supplier No. L0637709

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2809-69-0
Molecular Formula C6H6
Molecular Weight (g/mol) 78.11
MDL Number MFCD00041617
InChI Key PCTCNWZFDASPLA-UHFFFAOYSA-N
Synonym 2,4-hexadiyne, dimethylbutadiyne, dimethyldiacetylene, acmc-209h0v, ch3c.$.cc.$.cch3, hexa-2,4-diyn-1-yl
PubChem CID 137727
IUPAC Name hexa-2,4-diyne
SMILES CC#CC#CC

Specifications

Melting Point 66°C to 68°C
Boiling Point 127°C to 129°C
Quantity 10 g
UN Number UN1325
Beilstein 1731159
Formula Weight 78.11
Percent Purity 98%
Chemical Name or Material 2,4-Hexadiyne
Hazard Category H228
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.