accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNHNHNN

CAS RN 28128-19-0

IUPAC Name: 3,7-dihydro-2H-purine-2-thione
Synonyms: 3,7-dihydropurine-2-thione
Molecular Weight (g/mol): 152.18
Molecular Formula: C5H4N4S
InChi Key: HDBQZGJWHMCXIL-UHFFFAOYSA-N
SMILES: S=C1NC2=C(NC=N2)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results