Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptopurine, MP Biomedicals™

Catalog No. ICN15535680 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15535680 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15535680 Supplier MP Biomedicals, Inc Supplier No. 0215535680

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28128-19-0
Molecular Formula C5H4N4S
Molecular Weight (g/mol) 152.18
MDL Number MFCD00047145
InChI Key HDBQZGJWHMCXIL-UHFFFAOYSA-N
Synonym 2-mercaptopurine, 2-thiopurine, 5-mercaptopurine, purine-2-thiol, 2-purinethiol, 7h-purine-2-thiol, 9h-purine-2-thiol, 2-mp, 1,3-dihydro-2h-purine-2-thione, 3,7-dihydro-2h-purine-2-thione
PubChem CID 3015569
IUPAC Name 3,7-dihydro-2H-purine-2-thione
SMILES S=C1NC2=C(NC=N2)C=N1

Specifications

Color Orange
Quantity 100 mg
Formula Weight 152.18
Physical Form Powder
Chemical Name or Material 2-Mercaptopurine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.