accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28134-31-8

IUPAC Name: 2-ethylbenzoic acid
Synonyms: 2-ethylbenzoic acid
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: CGMMPMYKMDITEA-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results