Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Ethylbenzoic Acid, MP Biomedicals

Catalog No. ICN224539 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN224539 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN224539 Supplier MP Biomedicals, Inc Supplier No. 0522453901

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28134-31-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
InChI Key CGMMPMYKMDITEA-UHFFFAOYSA-N
Synonym benzoic acid, ethyl, o-ethylbenzoic acid, 2-ethylbenzoicacidethylester, ethylbenzoic acid, pubchem9869, 2-ethyl-benzoic acid, benzoic acid,2-ethyl, benzoic acid, 2-ethyl, acmc-1ax4g, 2-ethylbenzoic acid
PubChem CID 34170
IUPAC Name 2-ethylbenzoic acid
SMILES CCC1=CC=CC=C1C(=O)O

Specifications

Quantity 1 g
Formula Weight 150.13
Chemical Name or Material o-Ethylbenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.