accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFCH3CH3H3CCH3NNCH3H3CCH3CH3

CAS RN 282109-83-5

IUPAC Name: 1,3-bis[2,6-bis(propan-2-yl)phenyl]-4,5-dihydro-3H-1λ⁵-imidazol-1-ylium; tetrafluoroboranuide
Synonyms: 1,3-bis(2,6-diisopropylphenyl)-4,5-dihydro-1λ⁵-imidazol-1-ylium tetrafluoroborate
Molecular Weight (g/mol): 478.43
Molecular Formula: C27H39BF4N2
InChi Key: KFZBJQUHHALFSR-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CC(C)C1=CC=CC(C(C)C)=C1N1CC[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results