CAS RN 2825-82-3
CAS RN 2825-82-3
IUPAC Name:
tricyclo[5.2.1.0²,⁶]decane
Synonyms:
tricyclo[5.2.1.0²,⁶]decane
Molecular Weight (g/mol):
136.24
Molecular Formula:
C10H16
InChi Key:
LPSXSORODABQKT-UHFFFAOYNA-N
SMILES:
C1CC2C3CCC(C3)C2C1
1
–
1
of
1
results
1
exo-Tetrahydrodicyclopentadiene 94.0+%, TCI America™
CAS: 2825-82-3 Molecular Formula: C10H16 Molecular Weight (g/mol): 136.24 MDL Number: MFCD01714931 InChI Key: LPSXSORODABQKT-UHFFFAOYNA-N Synonym: exo-Tricyclo[5.2.1.0(2,6)]decane PubChem CID: 11159354 IUPAC Name: tricyclo[5.2.1.0²,⁶]decane SMILES: C1CC2C3CCC(C3)C2C1