accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONCH3OO

CAS RN 282526-98-1

IUPAC Name: 2-(hexadecyloxy)-6-methyl-4H-3,1-benzoxazin-4-one
Synonyms: cetilistat
Molecular Weight (g/mol): 401.59
Molecular Formula: C25H39NO3
InChi Key: MVCQKIKWYUURMU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCOC1=NC2=CC=C(C)C=C2C(=O)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results