CAS RN 2826-26-8
IUPAC Name:
2-[(4-methoxyphenyl)methylidene]propanedinitrile
Synonyms:
2-[(4-methoxyphenyl)methylidene]propanedinitrile
Molecular Weight (g/mol):
184.2
Molecular Formula:
C11H8N2O
InChi Key:
UOHFCPXBKJPCAD-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C(C#N)C#N)C=C1
1
–
2
of
2
results
1
Tyrphostin A1 98.0+%, TCI America™
Tyrphostin A1 98.0+%, TCI America™ available now.
2
MilliporeSigma™ Calbiochem™ AG 9
An inactive compound that can be used as a negative control for inhibition of EGFR (IC50 >1250 μM for EGFR kinase).