Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ AG 9

Catalog No. 6583905MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-839-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-839-05MG Supplier MilliporeSigma™ Supplier No. 6583905MG

An inactive compound that can be used as a negative control for inhibition of EGFR (IC50 >1250 μM for EGFR kinase).

An inactive compound that can be used as a negative control for inhibition of EGFR (IC50 >1250μM for EGFR kinase). Has been shown to inhibit IL-2 stimulated Tyk-2 phosphorylation in ConA-activated T cells.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 98% by TLC
Target Proteins negative control for inhibition of EGFR
CAS 2826-26-8
Molecular Formula C11H8N2O
Formula Weight 184.2g/mol
Solubility Acetic acid, DMSO, or EtOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.