accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28277-59-0

IUPAC Name: cobalt(3+) tris(1,10-phenanthroline) tris(hexafluoro-λ⁵-phosphanuide)
Synonyms: cobalt(3+) tris(phen) trihexafluorophosphate
Molecular Weight (g/mol): 1034.46
Molecular Formula: C36H24CoF18N6P3
InChi Key: JYGNLHURFRGGRD-UHFFFAOYSA-N
SMILES: [Co+3].F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results