Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(1,10-phenanthroline)cobalt(III) Tris(hexafluorophosphate) 97.0+%, TCI America™
SDP

Catalog No. T35545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35545G 5 g
T35541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35545G Supplier TCI America Supplier No. T35545G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28277-59-0
Molecular Formula C36H24CoF18N6P3
Molecular Weight (g/mol) 1034.46
MDL Number MFCD30749311
InChI Key JYGNLHURFRGGRD-UHFFFAOYSA-N
PubChem CID 15494322
IUPAC Name cobalt(3+) tris(1,10-phenanthroline) tris(hexafluoro-λ⁵-phosphanuide)
SMILES [Co+3].F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Quantity 5 g
Formula Weight 1034.46
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(1,10-phenanthroline)cobalt(III) Tris(hexafluorophosphate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.