CAS RN 28300-74-5
Antimony potassium tartrate trihydrate, ACS, 99.0-103.0%
CAS: 28300-74-5 Molecular Formula: C8H4K2O12Sb2·3H2O MDL Number: MFCD00148863 Synonym: Potassium antimonyl tartrate
Potassium antimonyl tartrate trihydrate, ACS reagent
CAS: 28300-74-5 Molecular Formula: 1·5 H2O Molecular Weight (g/mol): 333.93 Synonym: Antimonyl potassium tartrate sesquihydrate
Potassium antimonyl tartrate trihydrate, 99+%
CAS: 28300-74-5 Molecular Formula: C8H10K2O15Sb2 Molecular Weight (g/mol): 667.87 MDL Number: MFCD00167056 Synonym: Antimonyl potassium tartrate sesquihydrate
Potassium Antimony Tartrate Trihydrate, ACS, 99-103%, Spectrum™ Chemical
CAS: 28300-74-5
Potassium Antimony Tartrate Trihydrate, ACS Reagent Grade, Ricca Chemical
CAS: 28300-74-5 Molecular Weight (g/mol): 671.9 g/mol
Potassium Antimony Tartrate, ACS Grade, 99.0 to 103.0%, LabChem™
CAS: 28300-74-5
Potassium Antimony Tartrate Trihydrate, ACS Reagent (Tartar Emetic), Reagents
CAS: 28300-74-5 Molecular Formula: C8H10K2O15Sb2 Molecular Weight (g/mol): 667.87 InChI Key: WBTCZEPSIIFINA-UHFFFAOYNA-J Synonym: Potassium Antimonyl Tartrate Trihydrate IUPAC Name: dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate trihydrate SMILES: O.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2
Sigma Aldrich Potassium Antimonyl Tartrate Trihydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Potassium Antimonyl Tartrate Trihydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Antimony Potassium Tartrate Trihydrate (USP), Fisher Chemical™
CAS: 28300-74-5 Molecular Formula: C8H4K2O12Sb2 · 3 H2O MDL Number: MFCD00167056 Synonym: Tartrated Antimony
Antimony Potassium Tartrate, Powder, USP, 99-103%, Spectrum™ Chemical
CAS: 28300-74-5 Molecular Formula: C8H10K2O15Sb2 Molecular Weight (g/mol): 667.87 InChI Key: WBTCZEPSIIFINA-UHFFFAOYNA-J IUPAC Name: dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate trihydrate SMILES: O.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2