Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony Potassium Tartrate, Powder, USP, 99-103%, Spectrum™ Chemical
SDP

Catalog No. 18600737 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-737 12 kg Poly Pail
18-600-735 2.5 kg Poly Pail
18-600-736 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-600-737 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AN14012KGBL

missing translation for 'validMainframeMsg'

AN140, 28300-74-5, C8H4K2O12Sb2·3H2O

Spectrum™ Chemical Antimony Potassium Tartrate, Powder, USP is also referred to as emetic tartar. It has long been known as a powerful emetic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 28300-74-5
Molecular Formula C8H10K2O15Sb2
Molecular Weight (g/mol) 667.87
InChI Key WBTCZEPSIIFINA-UHFFFAOYNA-J
IUPAC Name dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate trihydrate
SMILES O.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2

Specifications

CAS Max % 100%
Quantity 12 kg
UN Number UN1551
Percent Purity 99 to 103%
Grade USP
Loss on Drying 2.7%
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.