CAS RN 2831-66-5
IUPAC Name:
2,4-dichloro-1,3,5-triazine
Synonyms:
2,4-dichloro-1,3,5-triazine
Molecular Weight (g/mol):
149.96
Molecular Formula:
C3HCl2N3
InChi Key:
OMRXVBREYFZQHU-UHFFFAOYSA-N
SMILES:
ClC1=NC(Cl)=NC=N1
1
–
1
of
1
results
1
2,4-Dichloro-1,3,5-triazine 98.0+%, TCI America™
CAS: 2831-66-5 Molecular Formula: C3HCl2N3 Molecular Weight (g/mol): 149.962 MDL Number: MFCD04115347 InChI Key: OMRXVBREYFZQHU-UHFFFAOYSA-N PubChem CID: 17810 IUPAC Name: 2,4-dichloro-1,3,5-triazine SMILES: C1=NC(=NC(=N1)Cl)Cl