accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28320-62-9

IUPAC Name: 9,9-dimethyl-9H-fluorene-2-carboxylic acid
Synonyms: 9,9-dimethylfluorene-2-carboxylic acid
Molecular Weight (g/mol): 238.29
Molecular Formula: C16H14O2
InChi Key: SJIBSIBHNJAUOR-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2C2=C1C=C(C=C2)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results