Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Dimethylfluorene-2-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. D33321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D33321G Supplier TCI America Supplier No. D33321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28320-62-9
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
InChI Key SJIBSIBHNJAUOR-UHFFFAOYSA-N
PubChem CID 44629795
IUPAC Name 9,9-dimethylfluorene-2-carboxylic acid
SMILES CC1(C2=CC=CC=C2C3=C1C=C(C=C3)C(=O)O)C

Specifications

Melting Point 196°C
Color White
Quantity 1 g
Formula Weight 238.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Dimethylfluorene-2-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.