accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2835-78-1

NH2O

CAS RN 2835-78-1

IUPAC Name: 3-benzoylaniline
Synonyms: benzophenone, 3-amino-
Molecular Weight (g/mol): 197.24
Molecular Formula: C13H11NO
InChi Key: FUADXEJBHCKVBN-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)