accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CON(E)

CAS RN 28446-68-6

IUPAC Name: (2E)-3-(4-methoxyphenyl)prop-2-enenitrile
Synonyms: cinnamonitrile, p-methoxy-
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: CNBCNHHPIBKYBO-NSCUHMNNSA-N
SMILES: COC1=CC=C(\C=C\C#N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results