Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxycinnamonitrile (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. M10335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10335G Supplier TCI America Supplier No. M10335G

Chemical Identifiers

CAS 28446-68-6
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00001934
InChI Key CNBCNHHPIBKYBO-NSCUHMNNSA-N
Synonym p-methoxycinnamonitrile, cinnamonitrile, p-methoxy, 4-methoxycinnamonitrile, 2-propenenitrile, 3-4-methoxyphenyl, cis,trans-4-methoxycinnamonitrile, 4-methoxycinnamonitrile,c&t, 4-methoxycinnamonitrile cis + trans, 2e-3-4-methoxyphenyl prop-2-enenitrile, 2-propenenitrile,3-4-methoxyphenyl, cambridge id 5106708
PubChem CID 1549677
IUPAC Name (E)-3-(4-methoxyphenyl)prop-2-enenitrile
SMILES COC1=CC=C(C=C1)C=CC#N

Specifications

Color Yellow
Quantity 5 g
Formula Weight 159.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Methoxycinnamonitrile (cis- and trans- mixture)
RTECSNumber GE1459320
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.