accessibility menu, dialog, popup

UserName

Viewing profile as user:
OCNSOO

CAS RN 2845-62-7

IUPAC Name: benzenesulfonyl isocyanate
Synonyms: benzenesulfonyl isocyanate
Molecular Weight (g/mol): 183.18
Molecular Formula: C7H5NO3S
InChi Key: UJYAZVSPFMJCLW-UHFFFAOYSA-N
SMILES: O=C=NS(=O)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results