Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzenesulfonyl Isocyanate 97.0+%, TCI America™
SDP

Catalog No. B16555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B16555G Supplier TCI America Supplier No. B16555G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2845-62-7
Molecular Formula C7H5NO3S
Molecular Weight (g/mol) 183.181
MDL Number MFCD00007546
InChI Key UJYAZVSPFMJCLW-UHFFFAOYSA-N
Synonym benzenesulfonyl isocyanate, phenylsulphonyl isocyanate, phenylsulfonylisocyanate, benzenesulfonylisocyanate, phenylsulfonyl isocyanate, benzenesulphonylisocyanate, phenyl sulfonyl isocyanate, acmc-1clf4, benzene sulfonyl isocyanate, phenyl sulphonyl isocyanate
PubChem CID 76096
IUPAC Name N-(oxomethylidene)benzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)N=C=O

Specifications

Color Yellow
Boiling Point 139°C
Quantity 5 g
UN Number 2206
Formula Weight 183.18
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Benzenesulfonyl Isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.