CAS RN 28485-18-9
CAS RN 28485-18-9
IUPAC Name:
2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride
Synonyms:
2,4-dioxo-1,3-dihydropyrimidine-5-sulfonyl chloride
Molecular Weight (g/mol):
210.59
Molecular Formula:
C4H3ClN2O4S
InChi Key:
FQHNJCWWBKYCJW-UHFFFAOYSA-N
SMILES:
ClS(=O)(=O)C1=CNC(=O)NC1=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals 2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride, 97%
CAS: 28485-18-9 Molecular Formula: C4H3ClN2O4S Molecular Weight (g/mol): 210.59 MDL Number: MFCD06655485 InChI Key: FQHNJCWWBKYCJW-UHFFFAOYSA-N Synonym: 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride,2,4-dioxo-1,3-dihydropyrimidine-5-sulfonyl chloride,1,2,3,4-tetrahydro-2,4-dioxo-5-pyrimidinesulfonyl chloride,5-pyrimidinesulfonylchloride, 1,2,3,4-tetrahydro-2,4-dioxo,uracil-5-sulphonyl chloride,5-chlorosulfonyl-1,3-dihydropyrimidine-2,4-dione,2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinesulfonyl chloride,2,4-dioxo-1,2,3,4-tetrahydro-pyrimidine-5-sulfonyl chloride PubChem CID: 234620 SMILES: ClS(=O)(=O)C1=CNC(=O)NC1=O