accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28495-72-9

IUPAC Name: 4-methyl-N'-(pentan-3-ylidene)benzene-1-sulfonohydrazide
Synonyms: 4-methyl-N'-(pentan-3-ylidene)benzenesulfonohydrazide
Molecular Weight (g/mol): 254.35
Molecular Formula: C12H18N2O2S
InChi Key: GUEMDXLIHCONOI-UHFFFAOYSA-N
SMILES: CCC(CC)=NNS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results